Assay Details
Gene Symbol: LOC5565207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649500.2 | XP_001649550.1 | 2 - 3 | 420 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5565207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649500.2 | XP_001649550.1 | 2 - 3 | 420 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() |