Assay Details
Gene Symbol: LOC5577190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846019.1 | XP_021701711.1 | 2 - 3 | 875 | 103 | |
XM_021846020.1 | XP_021701712.1 | 2 - 3 | 875 | 103 |

More Information
Set Membership: |
![]() ![]() |
