Assay Details
Gene Symbol: LOC5578886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001664044.2 | XP_001664094.1 | 2 - 3 | 925 | 62 | |
XM_021839620.1 | XP_021695312.1 | 3 - 4 | 860 | 62 | ||
XM_021839625.1 | XP_021695317.1 | 3 - 4 | 701 | 62 | ||
XM_021839629.1 | XP_021695321.1 | 3 - 4 | 661 | 62 |

More Information
Set Membership: |
![]() ![]() |
