Assay Details
Gene Symbol: LOC5578886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001664044.2 | XP_001664094.1 | 1 - 2 | 812 | 97 | |
XM_021839620.1 | XP_021695312.1 | 2 - 3 | 747 | 97 | ||
XM_021839625.1 | XP_021695317.1 | 2 - 3 | 588 | 97 | ||
XM_021839629.1 | XP_021695321.1 | 2 - 3 | 548 | 97 |
