Assay Details
Gene Symbol: LOC5566155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850555.1 | XP_021706247.1 | 1 - 2 | 280 | 89 | |
XM_021850556.1 | XP_021706248.1 | 2 - 3 | 285 | 89 | ||
XM_021850557.1 | XP_021706249.1 | 1 - 2 | 278 | 89 | ||
XM_021850558.1 | XP_021706250.1 | 2 - 3 | 285 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
