Assay Details
Gene Symbol: LOC110674164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838046.1 | XP_021693738.1 | 6 - 7 | 1869 | 91 | |
XM_021838047.1 | XP_021693739.1 | 6 - 7 | 1892 | 91 | ||
XM_021838048.1 | XP_021693740.1 | 5 - 6 | 1759 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
