Assay Details
Gene Symbol: LOC110674164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838046.1 | XP_021693738.1 | 5 - 6 | 1464 | 82 | |
XM_021838047.1 | XP_021693739.1 | 5 - 6 | 1487 | 82 | ||
XM_021838048.1 | XP_021693740.1 | 4 - 5 | 1354 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
