Assay Details
Gene Symbol: LOC110674164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838046.1 | XP_021693738.1 | 2 - 3 | 961 | 61 | |
XM_021838047.1 | XP_021693739.1 | 2 - 3 | 984 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
