Assay Details
Gene Symbol: LOC5570347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843773.1 | XP_021699465.1 | 1 - 2 | 1055 | 61 | |
XM_021843774.1 | XP_021699466.1 | 2 - 3 | 961 | 61 | ||
XM_021843775.1 | XP_021699467.1 | 2 - 3 | 774 | 61 | ||
XM_021843776.1 | XP_021699468.1 | 1 - 2 | 1056 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021843773.1, XM_021843774.1, XM_021843775.1, XM_021843776.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
