Assay Details
Gene Symbol: LOC5570347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653126.2 | XP_001653176.2 | 4 - 5 | 900 | 124 | |
XM_021843773.1 | XP_021699465.1 | 5 - 6 | 2286 | 124 | ||
XM_021843774.1 | XP_021699466.1 | 6 - 7 | 2192 | 124 | ||
XM_021843775.1 | XP_021699467.1 | 6 - 7 | 2005 | 124 | ||
XM_021843776.1 | XP_021699468.1 | 5 - 6 | 2287 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
