Assay Details
Gene Symbol: LOC5566347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845078.1 | XP_021700770.1 | 16 - 17 | 2910 | 71 | |
XM_021845079.1 | XP_021700771.1 | 16 - 17 | 2866 | 71 | ||
XM_021845080.1 | XP_021700772.1 | 15 - 16 | 2542 | 71 |
