Assay Details
Gene Symbol: LOC5570003 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849900.1 | XP_021705592.1 | 11 - 12 | 3544 | 73 | |
XM_021849901.1 | XP_021705593.1 | 11 - 12 | 3526 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
