Assay Details
Gene Symbol: LOC5568923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847502.1 | XP_021703194.1 | 10 - 11 | 4172 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5568923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847502.1 | XP_021703194.1 | 10 - 11 | 4172 | 74 |
Set Membership: |
![]() ![]() ![]() ![]() |