Assay Details
Gene Symbol: LOC5568923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847502.1 | XP_021703194.1 | 7 - 8 | 3314 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5568923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847502.1 | XP_021703194.1 | 7 - 8 | 3314 | 79 |
Set Membership: |
![]() ![]() ![]() |