Assay Details
Gene Symbol: LOC5566058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650374.2 | XP_001650424.1 | 1 - 2 | 924 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001650374.2 transcripts.
Set Membership: |
![]() ![]() |
