Assay Details
Gene Symbol: LOC5566058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650374.2 | XP_001650424.1 | 3 - 4 | 1618 | 92 | |
XM_021853165.1 | XP_021708857.1 | 2 - 3 | 821 | 92 | ||
XM_021853166.1 | XP_021708858.1 | 2 - 3 | 831 | 92 |
