Assay Details
Gene Symbol: LOC5575304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852830.1 | XP_021708522.1 | 2 - 3 | 413 | 109 | |
XM_021852831.1 | XP_021708523.1 | 2 - 3 | 277 | 109 | ||
XM_021852833.1 | XP_021708525.1 | 1 - 2 | 1085 | 109 | ||
XM_021852834.1 | XP_021708526.1 | 1 - 2 | 1086 | 109 | ||
XM_021852835.1 | XP_021708527.1 | 1 - 2 | 1085 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021852830.1, XM_021852831.1, XM_021852833.1, XM_021852834.1, XM_021852835.1 transcripts.
Set Membership: |
![]() ![]() |
