Assay Details
Gene Symbol: LOC5575304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852830.1 | XP_021708522.1 | 7 - 8 | 1747 | 77 | |
XM_021852831.1 | XP_021708523.1 | 7 - 8 | 1611 | 77 | ||
XM_021852832.1 | XP_021708524.1 | 6 - 7 | 1502 | 77 | ||
XM_021852833.1 | XP_021708525.1 | 5 - 6 | 2407 | 77 | ||
XM_021852834.1 | XP_021708526.1 | 6 - 7 | 2408 | 77 | ||
XM_021852835.1 | XP_021708527.1 | 6 - 7 | 2419 | 77 | ||
XM_021852836.1 | XP_021708528.1 | 3 - 4 | 1287 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
