Assay Details
Gene Symbol: LOC5575133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842205.1 | XP_021697897.1 | 2 - 3 | 397 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021842205.1 transcripts.
Set Membership: |
![]() ![]() |
