Assay Details
Gene Symbol: LOC5574473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655082.2 | XP_001655132.1 | 3 - 4 | 1183 | 105 | |
XM_021843927.1 | XP_021699619.1 | 3 - 4 | 1183 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
