Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 11 - 12 | 2306 | 101 | |
XM_021847847.1 | XP_021703539.1 | 11 - 12 | 2300 | 101 | ||
XM_021847848.1 | XP_021703540.1 | 11 - 12 | 2043 | 101 | ||
XM_021847849.1 | XP_021703541.1 | 11 - 12 | 1932 | 101 | ||
XM_021847850.1 | XP_021703542.1 | 15 - 16 | 3644 | 101 | ||
XM_021847851.1 | XP_021703543.1 | 14 - 15 | 3506 | 101 | ||
XM_021847852.1 | XP_021703544.1 | 13 - 14 | 3377 | 101 | ||
XM_021847853.1 | XP_021703545.1 | 5 - 6 | 1749 | 101 |

More Information
Set Membership: |
![]() ![]() |
