Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 12 - 13 | 2721 | 107 | |
XM_021847847.1 | XP_021703539.1 | 12 - 13 | 2715 | 107 | ||
XM_021847848.1 | XP_021703540.1 | 12 - 13 | 2458 | 107 | ||
XM_021847849.1 | XP_021703541.1 | 12 - 13 | 2347 | 107 | ||
XM_021847850.1 | XP_021703542.1 | 16 - 17 | 4059 | 107 | ||
XM_021847851.1 | XP_021703543.1 | 15 - 16 | 3921 | 107 | ||
XM_021847852.1 | XP_021703544.1 | 14 - 15 | 3792 | 107 | ||
XM_021847853.1 | XP_021703545.1 | 6 - 7 | 2164 | 107 |

More Information
Set Membership: |
![]() ![]() |
