Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 13 - 14 | 2857 | 104 | |
XM_021847847.1 | XP_021703539.1 | 13 - 14 | 2851 | 104 | ||
XM_021847848.1 | XP_021703540.1 | 13 - 14 | 2594 | 104 | ||
XM_021847849.1 | XP_021703541.1 | 13 - 14 | 2483 | 104 | ||
XM_021847850.1 | XP_021703542.1 | 17 - 18 | 4195 | 104 | ||
XM_021847851.1 | XP_021703543.1 | 16 - 17 | 4057 | 104 | ||
XM_021847852.1 | XP_021703544.1 | 15 - 16 | 3928 | 104 | ||
XM_021847853.1 | XP_021703545.1 | 7 - 8 | 2300 | 104 |

More Information
Set Membership: |
![]() ![]() |
