Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 14 - 15 | 3000 | 88 | |
XM_021847847.1 | XP_021703539.1 | 14 - 15 | 2994 | 88 | ||
XM_021847848.1 | XP_021703540.1 | 14 - 15 | 2737 | 88 | ||
XM_021847849.1 | XP_021703541.1 | 14 - 15 | 2626 | 88 | ||
XM_021847850.1 | XP_021703542.1 | 18 - 19 | 4338 | 88 | ||
XM_021847851.1 | XP_021703543.1 | 17 - 18 | 4200 | 88 | ||
XM_021847852.1 | XP_021703544.1 | 16 - 17 | 4071 | 88 | ||
XM_021847853.1 | XP_021703545.1 | 8 - 9 | 2443 | 88 |
