Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 15 - 16 | 4517 | 99 | |
XM_021847847.1 | XP_021703539.1 | 15 - 16 | 4511 | 99 | ||
XM_021847848.1 | XP_021703540.1 | 15 - 16 | 4254 | 99 | ||
XM_021847849.1 | XP_021703541.1 | 15 - 16 | 4143 | 99 | ||
XM_021847850.1 | XP_021703542.1 | 19 - 20 | 5855 | 99 | ||
XM_021847851.1 | XP_021703543.1 | 18 - 19 | 5717 | 99 | ||
XM_021847852.1 | XP_021703544.1 | 17 - 18 | 5588 | 99 | ||
XM_021847853.1 | XP_021703545.1 | 9 - 10 | 3960 | 99 |
