Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 19 - 20 | 5340 | 101 | |
XM_021847847.1 | XP_021703539.1 | 19 - 20 | 5334 | 101 | ||
XM_021847848.1 | XP_021703540.1 | 19 - 20 | 5077 | 101 | ||
XM_021847849.1 | XP_021703541.1 | 19 - 20 | 4966 | 101 | ||
XM_021847850.1 | XP_021703542.1 | 23 - 24 | 6678 | 101 | ||
XM_021847851.1 | XP_021703543.1 | 22 - 23 | 6540 | 101 | ||
XM_021847852.1 | XP_021703544.1 | 21 - 22 | 6411 | 101 | ||
XM_021847853.1 | XP_021703545.1 | 13 - 14 | 4783 | 101 |

More Information
Set Membership: |
![]() ![]() |
