Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 24 - 25 | 6321 | 81 | |
XM_021847847.1 | XP_021703539.1 | 24 - 25 | 6315 | 81 | ||
XM_021847848.1 | XP_021703540.1 | 24 - 25 | 6058 | 81 | ||
XM_021847849.1 | XP_021703541.1 | 24 - 25 | 5947 | 81 | ||
XM_021847850.1 | XP_021703542.1 | 28 - 29 | 7659 | 81 | ||
XM_021847851.1 | XP_021703543.1 | 27 - 28 | 7521 | 81 | ||
XM_021847852.1 | XP_021703544.1 | 26 - 27 | 7392 | 81 | ||
XM_021847853.1 | XP_021703545.1 | 18 - 19 | 5764 | 81 |
