Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 30 - 31 | 7457 | 98 | |
XM_021847847.1 | XP_021703539.1 | 30 - 31 | 7451 | 98 | ||
XM_021847848.1 | XP_021703540.1 | 30 - 31 | 7194 | 98 | ||
XM_021847849.1 | XP_021703541.1 | 30 - 31 | 7083 | 98 | ||
XM_021847850.1 | XP_021703542.1 | 34 - 35 | 8795 | 98 | ||
XM_021847851.1 | XP_021703543.1 | 33 - 34 | 8657 | 98 | ||
XM_021847852.1 | XP_021703544.1 | 32 - 33 | 8528 | 98 | ||
XM_021847853.1 | XP_021703545.1 | 24 - 25 | 6900 | 98 |
