Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 31 - 32 | 7886 | 71 | |
XM_021847847.1 | XP_021703539.1 | 31 - 32 | 7880 | 71 | ||
XM_021847848.1 | XP_021703540.1 | 31 - 32 | 7623 | 71 | ||
XM_021847849.1 | XP_021703541.1 | 31 - 32 | 7512 | 71 | ||
XM_021847850.1 | XP_021703542.1 | 35 - 36 | 9224 | 71 | ||
XM_021847851.1 | XP_021703543.1 | 34 - 35 | 9086 | 71 | ||
XM_021847852.1 | XP_021703544.1 | 33 - 34 | 8957 | 71 | ||
XM_021847853.1 | XP_021703545.1 | 25 - 26 | 7329 | 71 |
