Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 33 - 34 | 9389 | 72 | |
XM_021847847.1 | XP_021703539.1 | 33 - 34 | 9383 | 72 | ||
XM_021847848.1 | XP_021703540.1 | 33 - 34 | 9126 | 72 | ||
XM_021847849.1 | XP_021703541.1 | 33 - 34 | 9015 | 72 | ||
XM_021847850.1 | XP_021703542.1 | 37 - 38 | 10727 | 72 | ||
XM_021847851.1 | XP_021703543.1 | 36 - 37 | 10589 | 72 | ||
XM_021847852.1 | XP_021703544.1 | 35 - 36 | 10460 | 72 | ||
XM_021847853.1 | XP_021703545.1 | 27 - 28 | 8832 | 72 |
