Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 2 - 3 | 618 | 79 | |
XM_021847847.1 | XP_021703539.1 | 2 - 3 | 612 | 79 | ||
XM_021847848.1 | XP_021703540.1 | 2 - 3 | 355 | 79 | ||
XM_021847849.1 | XP_021703541.1 | 2 - 3 | 244 | 79 | ||
XM_021847850.1 | XP_021703542.1 | 6 - 7 | 1974 | 79 | ||
XM_021847851.1 | XP_021703543.1 | 6 - 7 | 1974 | 79 | ||
XM_021847852.1 | XP_021703544.1 | 6 - 7 | 1974 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021847846.1, XM_021847847.1, XM_021847848.1, XM_021847849.1, XM_021847850.1, XM_021847851.1, XM_021847852.1 transcripts.
Set Membership: |
![]() ![]() |
