Assay Details
Gene Symbol: LOC5579975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841705.1 | XP_021697397.1 | 1 - 2 | 167 | 113 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021841705.1 transcripts.
Set Membership: |
![]() ![]() |
