Assay Details
Gene Symbol: LOC5575240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655595.2 | XP_001655645.1 | 1 - 2 | 225 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001655595.2 transcripts.
Set Membership: |
![]() ![]() |
