Assay Details
Gene Symbol: LOC5574477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843935.1 | XP_021699627.1 | 11 - 12 | 2788 | 62 | |
XM_021843936.1 | XP_021699628.1 | 11 - 12 | 2783 | 62 | ||
XM_021843937.1 | XP_021699629.1 | 11 - 12 | 2770 | 62 | ||
XM_021843938.1 | XP_021699630.1 | 11 - 12 | 2765 | 62 | ||
XM_021843939.1 | XP_021699631.1 | 10 - 11 | 2251 | 62 | ||
XM_021843940.1 | XP_021699632.1 | 10 - 11 | 2281 | 62 | ||
XM_021843941.1 | XP_021699633.1 | 8 - 9 | 1388 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
