Assay Details
Gene Symbol: LOC5574477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843935.1 | XP_021699627.1 | 4 - 5 | 1594 | 69 | |
XM_021843936.1 | XP_021699628.1 | 4 - 5 | 1589 | 69 | ||
XM_021843937.1 | XP_021699629.1 | 4 - 5 | 1576 | 69 | ||
XM_021843938.1 | XP_021699630.1 | 4 - 5 | 1571 | 69 | ||
XM_021843939.1 | XP_021699631.1 | 3 - 4 | 1057 | 69 | ||
XM_021843940.1 | XP_021699632.1 | 3 - 4 | 1087 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
