Assay Details
Gene Symbol: LOC110678676 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851882.1 | XP_021707574.1 | 3 - 4 | 1466 | 96 | |
XM_021851883.1 | XP_021707575.1 | 3 - 4 | 1463 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
