Assay Details
Gene Symbol: LOC5576115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662411.2 | XP_001662461.2 | 1 - 2 | 63 | 76 | |
XM_021838398.1 | XP_021694090.1 | 2 - 3 | 313 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001662411.2, XM_021838398.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
