Assay Details
Gene Symbol: LOC5580263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854751.1 | XP_021710443.1 | 2 - 3 | 1416 | 64 | |
XM_021854752.1 | XP_021710444.1 | 2 - 3 | 1397 | 64 | ||
XM_021854753.1 | XP_021710445.1 | 2 - 3 | 1303 | 64 | ||
XM_021854754.1 | XP_021710446.1 | 2 - 3 | 1373 | 64 | ||
XM_021854755.1 | XP_021710447.1 | 2 - 3 | 1234 | 64 | ||
XM_021854756.1 | XP_021710448.1 | 2 - 3 | 1207 | 64 | ||
XM_021854757.1 | XP_021710449.1 | 2 - 3 | 1129 | 64 | ||
XM_021854759.1 | XP_021710451.1 | 2 - 3 | 1047 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
