Assay Details
Gene Symbol: LOC5577425 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663304.3 | XP_001663354.1 | 1 | 121 | ||
GenBank mRNA | AY432175.1 | - | 1 | 321 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
