Assay Details
Gene Symbol: LOC5578276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663754.2 | XP_001663804.1 | 4 - 5 | 894 | 104 | |
GenBank mRNA | AY432204.1 | - | 3 - 4 | 644 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
