Assay Details
Gene Symbol: LOC5577499 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656464.2 | XP_001656514.2 | 2 - 3 | 1006 | 71 | |
XM_001656465.2 | XP_001656515.2 | 2 - 3 | 1006 | 71 | ||
GenBank mRNA | AY432207.1 | - | 1 - 2 | 277 | 71 | |
DQ440278.1 | - | 2 - 3 | 743 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
