Assay Details
Gene Symbol: LOC5577499 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656465.2 | XP_001656515.2 | 3 - 4 | 1110 | 90 | |
GenBank mRNA | AY432207.1 | - | 2 - 3 | 381 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
