Assay Details
Gene Symbol: LOC5574338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661321.2 | XP_001661371.1 | 2 - 3 | 499 | 108 | |
GenBank mRNA | AY432456.1 | - | 2 - 3 | 475 | 108 | |
DQ440308.1 | - | 2 - 3 | 357 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
