Assay Details
Gene Symbol: LOC5575432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655700.2 | XP_001655750.2 | 2 - 3 | 218 | 87 | |
XM_001655702.2 | XP_001655752.2 | 2 - 3 | 218 | 87 | ||
XM_011494673.2 | XP_011492975.2 | 2 - 3 | 116 | 87 |
