Assay Details
Gene Symbol: LOC5575432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655700.2 | XP_001655750.2 | 5 - 6 | 481 | 85 | |
XM_001655702.2 | XP_001655752.2 | 4 - 5 | 373 | 85 | ||
XM_001655705.2 | XP_001655755.2 | 4 - 5 | 341 | 85 | ||
XM_011494673.2 | XP_011492975.2 | 5 - 6 | 379 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
