Assay Details
Gene Symbol: LOC5568156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843214.1 | XP_021698906.1 | 3 - 4 | 2209 | 68 | |
XM_021843215.1 | XP_021698907.1 | 4 - 5 | 2174 | 68 |

More Information
Set Membership: |
![]() ![]() |
