Assay Details
Gene Symbol: LOC5568156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843214.1 | XP_021698906.1 | 4 - 5 | 3302 | 84 | |
XM_021843215.1 | XP_021698907.1 | 5 - 6 | 3267 | 84 |
