Assay Details
Gene Symbol: LOC5569229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857565.1 | XP_021713257.1 | 1 - 2 | 89 | 57 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021857565.1 transcripts.
Set Membership: |
![]() ![]() |
