Assay Details
Gene Symbol: LOC5569062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847326.1 | XP_021703018.1 | 7 - 8 | 6006 | 60 | |
XM_021847327.1 | XP_021703019.1 | 7 - 8 | 5385 | 60 | ||
XM_021847328.1 | XP_021703020.1 | 7 - 8 | 6006 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
