Assay Details
Gene Symbol: LOC5572212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653830.2 | XP_001653880.2 | 3 - 4 | 3153 | 58 | |
XM_021843085.1 | XP_021698777.1 | 4 - 5 | 2965 | 58 | ||
XM_021843086.1 | XP_021698778.1 | 4 - 5 | 2919 | 58 | ||
XM_021843087.1 | XP_021698779.1 | 3 - 4 | 3153 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
